Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081141.1 | NP_001074610.1 | 1 - 2 | 608 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |