Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009752.2 | NP_033882.1 | 3 - 4 | 515 | 81 | |
NR_108101.1 | 2 - 3 | 345 | 81 | |||
NR_108102.1 | 2 - 3 | 345 | 81 | |||
GenBank mRNA | AK034049.1 | - | 3 - 4 | 494 | 81 | |
AK034966.1 | - | 316 | 81 | |||
AK042069.1 | - | 3 - 4 | 485 | 81 | ||
AK048460.1 | - | 3 - 4 | 446 | 81 | ||
AK076517.1 | - | 3 - 4 | 486 | 81 | ||
AK153218.1 | - | 3 - 4 | 498 | 81 | ||
AK166473.1 | - | 3 - 4 | 500 | 81 | ||
AK171593.1 | - | 3 - 4 | 518 | 81 | ||
AK171760.1 | - | 2 - 3 | 316 | 81 | ||
BC003998.1 | - | 1 - 2 | 86 | 81 | ||
BC028875.1 | - | 3 - 4 | 435 | 4240285 | 81 | |
M57734.1 | - | 3 - 4 | 474 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |