Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028947.1 | NP_083223.1 | 7 - 8 | 1022 | 101 | |
GenBank mRNA | AK016872.1 | - | 7 - 8 | 1023 | 101 | |
BC100522.1 | - | 7 - 8 | 920 | 30920038 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |