Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133858.4 | NP_598619.2 | 6 - 7 | 1832 | 96 | |
NM_199475.1 | NP_955769.1 | 6 - 7 | 1832 | 96 | ||
GenBank mRNA | AB075603.1 | - | 5 - 6 | 843 | 96 | |
AK036838.1 | - | 6 - 7 | 1824 | 96 | ||
AK087859.1 | - | 6 - 7 | 1645 | 96 | ||
AK169673.1 | - | 6 - 7 | 1832 | 96 | ||
AK182185.1 | - | 1 - 2 | 99 | 96 | ||
AK186674.1 | - | 1 - 2 | 99 | 96 | ||
AK215644.1 | - | 1 - 2 | 99 | 96 | ||
BC039762.1 | - | 5 - 6 | 650 | 96 | ||
BC051048.1 | - | 5 - 6 | 715 | 96 | ||
BC057901.1 | - | 4 - 5 | 361 | 96 | ||
BC093521.1 | - | 2 - 3 | 401 | 4007022 | 96 | |
BC094388.1 | - | 6 - 7 | 1649 | 5708548 | 96 | |
BC132301.1 | - | 5 - 6 | 1077 | 40130578 | 96 | |
BC144749.1 | - | 5 - 6 | 1077 | 9053297 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |