Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010664.2 | NP_034794.2 | 3 - 4 | 695 | 78 | |
GenBank mRNA | AK167072.1 | - | 3 - 4 | 684 | 78 | |
AK167432.1 | - | 3 - 4 | 682 | 78 | ||
AK167469.1 | - | 3 - 4 | 685 | 78 | ||
AK167676.1 | - | 3 - 4 | 684 | 78 | ||
BC020474.1 | - | 3 - 4 | 685 | 78 | ||
BC089022.1 | - | 3 - 4 | 657 | 30701486 | 78 | |
CV561123.1 | - | 3 - 4 | 697 | 78 | ||
M11686.1 | - | 3 - 4 | 686 | 78 | ||
M36376.1 | - | 3 - 4 | 681 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |