Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181732.4 | NP_859421.1 | 1 - 2 | 573 | 88 | |
GenBank mRNA | AK076031.1 | - | 1 - 2 | 225 | 88 | |
AK081494.1 | - | 1 - 2 | 574 | 88 | ||
AK143716.1 | - | 1 - 2 | 113 | 88 | ||
AK149788.1 | - | 1 - 2 | 325 | 88 | ||
AK150639.1 | - | 1 - 2 | 261 | 88 | ||
AK150702.1 | - | 1 - 2 | 322 | 88 | ||
AK151783.1 | - | 1 - 2 | 264 | 88 | ||
AK152961.1 | - | 1 - 2 | 264 | 88 | ||
AK153448.1 | - | 1 - 2 | 261 | 88 | ||
AK158103.1 | - | 1 - 2 | 207 | 88 | ||
AK159318.1 | - | 1 - 2 | 585 | 88 | ||
AK167520.1 | - | 1 - 2 | 290 | 88 | ||
AK172451.1 | - | 1 - 2 | 266 | 88 | ||
BC057183.1 | - | 1 - 2 | 228 | 5065689 | 88 | |
BC066829.1 | - | 1 - 2 | 112 | 88 | ||
BC086763.1 | - | 1 - 2 | 278 | 5707830 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |