Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
TC0500001025.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204201.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204201.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204202.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204202.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204203.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204203.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_001204233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_009263.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008647.mm.1 |
NM_009263.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008642.mm.1 |
TC0500001025.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008642.mm.1 |
NM_001204233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008642.mm.1 |
NM_001204233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008644.mm.1 |
TC0500001025.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008644.mm.1 |
NM_001204233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008644.mm.1 |
NM_001204233.1 |
Refseq |