Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700008767.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008767.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008767.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008767.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008767.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008790.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008790.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008790.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008793.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008793.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008793.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008793.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008793.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008811.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008799.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008809.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008773.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008773.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008773.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008773.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008773.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008777.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008777.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008777.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008777.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008777.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008796.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008812.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008818.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008789.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008789.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008789.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008782.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008782.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008782.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008782.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008782.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008817.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008807.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008813.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008768.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008768.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008768.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008768.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008768.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008795.mm.1 |
TC1700000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_001081635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_001081635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_001081636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_001081636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_007632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
NM_007632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008808.mm.1 |
TC1700000917.mm.1 |
MTA TC |