Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pydc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177349.1 | NP_001170820.1 | 4 | 1212 | 69 | |
NM_001177350.1 | NP_001170821.1 | 3 | 1160 | 69 | ||
GenBank mRNA | AK143015.1 | - | 4 | 1213 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon |