Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024427.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024427.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024427.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024429.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024429.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024429.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024430.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024430.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024430.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024433.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024433.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024433.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024434.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024434.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024434.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024440.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024440.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024440.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024438.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024438.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024438.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024432.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024432.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024432.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024439.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024439.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024439.mm.1 |
NM_017367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024431.mm.1 |
TC0500002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024431.mm.1 |
NM_017367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024431.mm.1 |
NM_017367.3 |
Refseq |