Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930519H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015844.1 | - | 1 - 2 | 238 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spermatogenesis associated glutamate (E)-rich protein 4c | Speer4c |
4933431D05Rik, SPEER-4C |
-155.4kb | Sense | Chr. 5 - 15709505 - 15714675 on NCBI Build GRCm38 |
Downstream | predicted gene 3510 | Gm3510 | 39.9kb | Sense | Chr. 5 - 15923754 - 15930127 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |