Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008855.2 | NP_032881.1 | 3 | 452 | 83 | |
GenBank mRNA | BC127083.1 | - | 3 | 271 | 40111744 | 83 |
BY729820.1 | - | 3 | 452 | 83 | ||
CJ050078.1 | - | 3 | 243 | 83 | ||
X53532.1 | - | 3 | 280 | 83 | ||
X59274.1 | - | 3 | 268 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Amplicon spans exons and probe does not span exons |