Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm36757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039743.1 | - | 2 | 3166 | 137 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 5033406O09 gene | 5033406O09Rik | 9130204G15Rik | -8.6kb | Anti-sense | Chr. 12 - 111941991 - 111944482 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 2010107E04 gene | 2010107E04Rik |
9430003J03Rik, AU043134, AV124504, Mp68 |
7.2kb | Anti-sense | Chr. 12 - 111961375 - 111966977 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |