Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412594 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412602 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412462 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412603 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412597 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412615 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412596 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412593 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412466 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412464 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412581 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412607 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412609 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412595 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412608 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412606 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412599 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412463 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412465 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412612 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412589 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412582 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412598 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412460 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412616 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412591 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412587 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412604 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412610 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412588 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412605 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412461 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412611 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412613 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963491 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957219 |
BC085171.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957219 |
X56135.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957219 |
AK159170.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957219 |
BC083135.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957219 |
BC083163.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2148746 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK088076.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK152693.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK160078.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK168160.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK168174.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK198109.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
AK209792.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
BC024855.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
BC025032.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
NM_146200.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
U67328.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563498 |
XM_001478697.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535081 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963491 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957219 |
BC085171.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957219 |
X56135.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957219 |
AK159170.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957219 |
BC083135.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957219 |
BC083163.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2148746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412568 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412576 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412558 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412561 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412566 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412573 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412569 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412556 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412557 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412578 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412565 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412602 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412579 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412586 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412574 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412588 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412564 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412562 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412559 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412620 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412589 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412585 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20412571 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00123524 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P249227 |
BC085171.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249227 |
X56135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249227 |
AK159170.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249227 |
BC083135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249227 |
BC083163.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P249402 |
BC085171.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P249402 |
X56135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P249402 |
BC081453.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P249402 |
BC083135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P249402 |
AK153953.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P107032 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00123524 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00844632 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00443803 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00443800 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00844651 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00844649 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412581 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412606 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412615 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412589 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412608 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412599 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412464 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412616 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412465 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412597 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412609 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412587 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412582 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412463 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412594 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412607 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412596 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412461 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412602 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412591 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412610 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412466 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412595 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412603 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412613 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412604 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412462 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412460 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412611 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412612 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412593 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412588 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412598 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20412605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412559 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20412612 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00593606 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01122362 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412652 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412617 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412573 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412653 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020827 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00844644 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412596 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00844653 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412551 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00844641 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412579 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589102 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00574772 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412549 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412607 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412555 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01122361 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412642 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00574769 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020825 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412583 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020815 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412589 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412554 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589100 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412574 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00443802 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00963090 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020817 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412556 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00593603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412626 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020822 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00844636 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412568 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00443803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412576 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00698149 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00443800 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00592721 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412594 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00565786 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01020823 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01122358 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412639 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589103 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589108 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412559 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00728469 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412553 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20412611 |
- |
- |