Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028014.mm.1 |
NM_030702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028014.mm.1 |
NM_030702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028014.mm.1 |
TC1100003058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028011.mm.1 |
NM_001163571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028011.mm.1 |
NM_001163571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028011.mm.1 |
NM_030702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028011.mm.1 |
NM_030702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028011.mm.1 |
TC1100003058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028017.mm.1 |
NM_001163571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028017.mm.1 |
NM_001163571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028017.mm.1 |
TC1100003058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028015.mm.1 |
NM_030702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028015.mm.1 |
NM_030702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028015.mm.1 |
TC1100003058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028013.mm.1 |
NM_001163571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028013.mm.1 |
NM_001163571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028013.mm.1 |
NM_030702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028013.mm.1 |
NM_030702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028013.mm.1 |
TC1100003058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028016.mm.1 |
NM_030702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028016.mm.1 |
NM_030702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028016.mm.1 |
TC1100003058.mm.1 |
MTA TC |