Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178786.4 | NP_848901.2 | 2 - 3 | 325 | 136 | |
GenBank mRNA | AK028795.1 | - | 2 - 3 | 326 | 136 | |
AK035748.1 | - | 2 - 3 | 243 | 136 | ||
EF494897.1 | - | 2 - 3 | 124 | 136 | ||
EF494898.1 | - | 2 - 3 | 124 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Probe spans exons |