Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207222.2 | NP_997105.1 | 3 - 4 | 403 | 108 | |
GenBank mRNA | AK034177.1 | - | 3 - 4 | 404 | 108 | |
BC034128.1 | - | 1 - 2 | 104 | 108 | ||
BC057086.1 | - | 2 - 3 | 639 | 5720992 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |