Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170673.3 | NP_733773.2 | 2 - 3 | 282 | 115 | |
GenBank mRNA | AK042692.1 | - | 2 - 3 | 283 | 115 | |
AK044080.1 | - | 2 - 3 | 248 | 115 | ||
AK047571.1 | - | 2 - 3 | 226 | 115 | ||
AK137422.1 | - | 2 - 3 | 283 | 115 | ||
BB639866.1 | - | 2 - 3 | 283 | 115 | ||
BC115728.1 | - | 2 - 3 | 232 | 40039295 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |