Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847845 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847848 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847846 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847847 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965045 |
AK133432.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965045 |
NM_001033542.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
434281 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623306 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647235 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
350565 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
276905 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
AK144842.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
AK160894.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
BC017545.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
BC072577.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
BC079851.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651249 |
NM_145607.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
443558 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
99620 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10222 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630699 |
AK081095.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946001 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
141967 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
730366 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334560 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
951849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602585 |
AK041776.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602585 |
AK042297.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
818396 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
566781 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57198 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925521 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611526 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3046 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
896910 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58575 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
68453 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
33177 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
140896 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965045 |
AK133432.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965045 |
NM_001033542.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847846 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847840 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847833 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847848 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847844 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27847847 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07364879 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07364833 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02340579 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07364804 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07364851 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P02340579 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07364833 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847847 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847846 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847845 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27847848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27847689 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09229654 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09551605 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847840 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847809 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847820 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847851 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09551617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847835 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847818 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847833 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847812 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09517360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09491810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09551616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09491636 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09517359 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08984374 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27847825 |
- |
- |