Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052835.3 | NP_443067.1 | 5 - 6 | 411 | 115 | |
GenBank mRNA | AK011022.1 | - | 5 - 6 | 371 | 115 | |
AK012561.1 | - | 4 - 5 | 368 | 115 | ||
AK014054.1 | - | 5 - 6 | 373 | 115 | ||
AK088777.1 | - | 5 - 6 | 372 | 115 | ||
AK144831.1 | - | 5 - 6 | 371 | 115 | ||
AK146217.1 | - | 5 - 6 | 372 | 115 | ||
AK151416.1 | - | 5 - 6 | 371 | 115 | ||
AK151655.1 | - | 5 - 6 | 371 | 115 | ||
AK151736.1 | - | 5 - 6 | 370 | 115 | ||
AK152123.1 | - | 5 - 6 | 371 | 115 | ||
AK152854.1 | - | 5 - 6 | 371 | 115 | ||
AK153345.1 | - | 5 - 6 | 371 | 115 | ||
AK160618.1 | - | 5 - 6 | 371 | 115 | ||
AK166610.1 | - | 5 - 6 | 370 | 115 | ||
AK167239.1 | - | 4 - 5 | 407 | 115 | ||
AK167966.1 | - | 5 - 6 | 371 | 115 | ||
AK168250.1 | - | 5 - 6 | 370 | 115 | ||
AK168854.1 | - | 5 - 6 | 371 | 115 | ||
AK187587.1 | - | 3 - 4 | 264 | 115 | ||
AK210081.1 | - | 3 - 4 | 260 | 115 | ||
BC006068.1 | - | 4 - 5 | 330 | 115 | ||
BC024901.1 | - | 4 - 5 | 332 | 4224016 | 115 | |
BC048872.1 | - | 4 - 5 | 335 | 5052784 | 115 | |
BC082293.1 | - | 4 - 5 | 332 | 6486491 | 115 | |
BC083327.1 | - | 4 - 5 | 332 | 5684015 | 115 | |
BC086917.1 | - | 5 - 6 | 370 | 6768088 | 115 | |
BC092383.1 | - | 4 - 5 | 356 | 5710462 | 115 | |
BC098204.1 | - | 4 - 5 | 356 | 6410755 | 115 | |
BC099437.1 | - | 4 - 5 | 335 | 30843551 | 115 | |
M93980.1 | - | 5 - 6 | 370 | 115 | ||
X75312.1 | - | 4 - 5 | 363 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |