Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008983.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008991.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008991.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008991.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008991.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008991.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008998.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008998.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008998.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008998.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008998.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008981.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008995.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008995.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008995.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008995.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008995.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008989.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008992.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008992.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008992.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008992.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008992.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008993.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008993.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008993.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008993.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008993.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008994.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008994.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008994.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008994.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008994.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008978.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008985.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008996.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008996.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008996.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008996.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008996.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008984.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008988.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008997.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008997.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008997.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008997.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008997.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
NR_033513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008980.mm.1 |
TC0200001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_053263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_053263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_146130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_146130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_198090.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
NM_198090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009001.mm.1 |
TC0200001115.mm.1 |
MTA TC |