Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100033662.mm.1 |
NM_001081435.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033662.mm.1 |
NM_001081435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033662.mm.1 |
TC1100003658.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033660.mm.1 |
NM_001081435.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033660.mm.1 |
NM_001081435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033660.mm.1 |
TC1100003658.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033659.mm.1 |
NM_001081435.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033659.mm.1 |
NM_001081435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033659.mm.1 |
TC1100003658.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033661.mm.1 |
NM_001081435.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033661.mm.1 |
NM_001081435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033661.mm.1 |
TC1100003658.mm.1 |
MTA TC |