Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026130.1 | NP_080406.1 | 9 - 10 | 1218 | 108 | |
GenBank mRNA | AK004967.2 | - | 9 - 10 | 1218 | 108 | |
AK038017.1 | - | 9 - 10 | 1206 | 108 | ||
AK177061.1 | - | 4 - 5 | 348 | 108 | ||
BC012512.1 | - | 301 | 108 | |||
BC021839.1 | - | 9 - 10 | 1201 | 4207633 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |