Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152824.1 | NP_690037.1 | 6 - 7 | 680 | 97 | |
GenBank mRNA | AF083384.1 | - | 5 - 6 | 550 | 97 | |
AK012296.1 | - | 6 - 7 | 626 | 97 | ||
BC034896.1 | - | 6 - 7 | 680 | 1446602 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |