Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800018132.mm.1 |
NM_001122830.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018132.mm.1 |
NM_001122830.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018132.mm.1 |
NM_178771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018132.mm.1 |
NM_178771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018132.mm.1 |
TC0800002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018128.mm.1 |
NM_001122830.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018128.mm.1 |
NM_001122830.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018128.mm.1 |
NM_178771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018128.mm.1 |
NM_178771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018128.mm.1 |
TC0800002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018131.mm.1 |
NM_001122830.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018131.mm.1 |
NM_001122830.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018131.mm.1 |
NM_178771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018131.mm.1 |
NM_178771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018131.mm.1 |
TC0800002392.mm.1 |
MTA TC |