Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinc | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ059756.1 | - | LIT3461 PINC |
5 - 6 | 731 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700027A15 gene | 1700027A15Rik | -369.8kb | Anti-sense | Chr. 1 - 73016036 - 73025508 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | 1.8kb | Anti-sense | Chr. 1 - 73398968 - 73430779 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |