Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa11os | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015348.1 | 3 - 4 | 1009 | 59 | |||
GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
3 - 4 | 1009 | 59 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox A11 | Hoxa11 |
Hox-1.9, Hoxa-11 |
-505.0 | Anti-sense | Chr. 6 - 52242105 - 52245810 on NCBI Build GRCm38 |
Downstream | homeobox A13 | Hoxa13 |
Hd, Hox-1.10 |
12.0kb | Anti-sense | Chr. 6 - 52258853 - 52260880 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |