Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278943.1 | NP_001265872.1 | 7 - 8 | 1131 | 70 | |
NM_001278944.1 | NP_001265873.1 | 10 - 11 | 1597 | 70 | ||
GenBank mRNA | AK144723.1 | - | 7 - 8 | 1132 | 70 | |
AY243472.1 | - | 10 - 11 | 1596 | 70 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATP-binding cassette, sub-family A (ABC1), member 15 |
Abca15 | 4930500I12Rik | -49.4kb | Sense | Chr. 7 - 120328486 - 120407687 on NCBI Build GRCm38 |
Downstream | Y box protein 1 pseudogene | E130201H02Rik | 131.8kb | Sense | Chr. 7 - 120597625 - 120598475 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |