Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278943.1 | NP_001265872.1 | 6 - 7 | 995 | 89 | |
NM_001278944.1 | NP_001265873.1 | 9 - 10 | 1461 | 89 | ||
GenBank mRNA | AK144723.1 | - | 6 - 7 | 996 | 89 | |
AY243472.1 | - | 9 - 10 | 1460 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATP-binding cassette, sub-family A (ABC1), member 15 |
Abca15 | 4930500I12Rik | -45.3kb | Sense | Chr. 7 - 120328486 - 120407687 on NCBI Build GRCm38 |
Downstream | Y box protein 1 pseudogene | E130201H02Rik | 140.4kb | Sense | Chr. 7 - 120597625 - 120598475 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons |