Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxl2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003248.3 | 4 - 5 | 1282 | 61 | ||
GenBank mRNA | AK143359.1 | - | 4 - 5 | 1272 | 61 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 7420426K07 gene | 7420426K07Rik | -47.7kb | Anti-sense | Chr. 9 - 98903119 - 98904622 on NCBI Build GRCm38 | |
Downstream | forkhead box L2 | Foxl2 |
AU045128, BPES, PINTO, Pfrk |
1.8kb | Anti-sense | Chr. 9 - 98955607 - 98958126 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |