Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007484.2 | NP_031510.2 | 1 - 2 | 46 | 74 | |
GenBank mRNA | AK152802.1 | - | 1 - 2 | 44 | 74 | |
AK160650.1 | - | 1 - 2 | 41 | 74 | ||
BY147599.1 | - | 1 - 2 | 47 | 74 | ||
X80638.1 | - | 1 - 2 | 28 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |