Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038266.1 | - | 1 | 251 | 88 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | trafficking protein particle complex 11 | Trappc11 |
AW558465, D030016E14Rik, R75422 |
-17.0kb | Anti-sense | Chr. 8 - 47490128 - 47533470 on NCBI Build GRCm38 |
Downstream |
inhibitor of growth family, member 2 |
Ing2 |
2810011M06Rik, Ing1l, Ing2b, P33ING2 |
114.2kb | Anti-sense | Chr. 8 - 47667178 - 47675609 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |