Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172467.3 | NP_766055.1 | 4 | 6108 | 110 | |
GenBank mRNA | AK045178.1 | - | 1 | 350 | 110 | |
AK053232.1 | - | 2 | 1082 | 110 | ||
AK088451.1 | - | 1 | 1459 | 110 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA D630045J12 gene | D630045J12Rik |
Kiaa1549, mKIAA1549 |
-33.4kb | Sense | Chr. 6 - 38123174 - 38254009 on NCBI Build GRCm38 |
Downstream |
zinc finger CCCH type, antiviral 1 |
Zc3hav1 |
1200014N16Rik, 2900058M19Rik, 9130009D18Rik, 9830115L13Rik, ARTD13, D6Bwg1452e, ZAP |
17.1kb | Sense | Chr. 6 - 38305286 - 38355840 on NCBI Build GRCm38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |