Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC022916.1 | - | 2 - 3 | 162 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BCL2/adenovirus E1B interacting protein 3-like | Bnip3l |
C86132, D14Ertd719e, Nip3L, Nix |
-53.5kb | Sense | Chr. 14 - 66985238 - 67008894 on NCBI Build GRCm38 |
Downstream | early B cell factor 2 | Ebf2 |
D14Ggc1e, EBF-2, Mmot1, O/E-3, OE-3 |
163.0kb | Anti-sense | Chr. 14 - 67233292 - 67430952 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |