Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172443.3 | NP_766031.2 | 12 | 6409 | 86 | |
GenBank mRNA | AK165823.1 | - | 1 | 2226 | 86 | |
BC026530.1 | - | 1 | 1050 | 86 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 11754 | Gm11754 | OTTMUSG00000004003 | -31.6kb | Sense | Chr. 11 - 119105130 - 119111450 on NCBI Build GRCm38 |
Downstream | microRNA 6934 | Mir6934 |
Gm27523, mmu-mir-6934 |
9.4kb | Sense | Chr. 11 - 119154076 - 119154137 on NCBI Build GRCm38 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |