Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012046.1 | - | 1 | 2047 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mediator complex subunit 15 | Med15 |
A230074L19Rik, AW536074, Pcqap, mPcqap |
-36.7kb | Anti-sense | Chr. 16 - 17651208 - 17722947 on NCBI Build GRCm38 |
Downstream |
scavenger receptor class F, member 2 |
Scarf2 |
AI194363, NSR1, SREC-II, SRECRP-1, Scarf1, Scatf1, Srec2, SrecII, W53987, W65560 |
35.4kb | Sense | Chr. 16 - 17797282 - 17808294 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |