Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm28722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132070.1 | - | 1 - 2 | 167 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102638195 | LOC102638195 | -162.6kb | Anti-sense | Chr. 1 - 90270711 - 90290770 on NCBI Build GRCm38 | |
Downstream |
COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana) |
Cops8 |
9430009J09Rik, AA408242, Csn8, Sgn8 |
148.9kb | Anti-sense | Chr. 1 - 90603425 - 90613341 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons |