Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100010263.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010263.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010263.mm.1 |
TC1100001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010265.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010265.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010265.mm.1 |
TC1100001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010262.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010262.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010262.mm.1 |
TC1100001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010261.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010261.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010261.mm.1 |
TC1100001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010264.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010264.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010264.mm.1 |
TC1100001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010266.mm.1 |
NM_001024205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010266.mm.1 |
NM_001024205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010266.mm.1 |
TC1100001131.mm.1 |
MTA TC |