Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159619.1 | NP_001153091.1 | 1 | 150 | 75 | |
NM_001159620.1 | NP_001153092.1 | 1 | 150 | 75 | ||
GenBank mRNA | AF216308.1 | - | 1 | 40 | 75 | |
BC061117.1 | - | 1 | 109 | 75 | ||
CB596599.1 | - | 1 | 153 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |