Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048168.2 | NP_001041633.1 | 2 - 3 | 256 | 104 | |
NM_001277095.1 | NP_001264024.1 | 2 - 3 | 329 | 104 | ||
NM_008692.4 | NP_032718.2 | 2 - 3 | 329 | 104 | ||
GenBank mRNA | AB006181.1 | - | 1 - 2 | 109 | 104 | |
AK011327.1 | - | 2 - 3 | 286 | 104 | ||
AK156353.1 | - | 2 - 3 | 183 | 104 | ||
BC020117.1 | - | 2 - 3 | 255 | 4009737 | 104 | |
BC053723.1 | - | 2 - 3 | 259 | 30053020 | 104 | |
BC085261.1 | - | 2 - 3 | 255 | 6397760 | 104 | |
CF172077.1 | - | 2 - 3 | 252 | 104 | ||
CJ092564.1 | - | 2 - 3 | 306 | 104 | ||
DV650959.1 | - | 2 - 3 | 268 | 104 | ||
U62297.1 | - | 1 - 2 | 109 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |