Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083925.1 | NP_001077394.1 | 2 - 3 | 521 | 111 | |
NR_003507.1 | 2 - 3 | 524 | 111 | |||
GenBank mRNA | AF328926.1 | - | 2 - 3 | 521 | 111 | |
AF418004.1 | - | 2 - 3 | 521 | 111 | ||
AF418005.1 | - | 2 - 3 | 521 | 111 | ||
AF418006.1 | - | 2 - 3 | 521 | 111 | ||
AF418007.1 | - | 2 - 3 | 521 | 111 | ||
AF418009.1 | - | 2 - 3 | 521 | 111 | ||
AF466822.1 | - | 2 - 3 | 478 | 111 | ||
AF466823.1 | - | 2 - 3 | 478 | 111 | ||
AK080152.1 | - | 2 - 3 | 514 | 111 | ||
BC012877.1 | - | 2 - 3 | 513 | 4016177 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |