Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004917.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004917.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004917.mm.1 |
TC1400000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004908.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004908.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004908.mm.1 |
TC1400000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004911.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004911.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004911.mm.1 |
TC1400000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004913.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004913.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004913.mm.1 |
TC1400000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004907.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004907.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004907.mm.1 |
TC1400000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004914.mm.1 |
NM_023434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004914.mm.1 |
NM_023434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004914.mm.1 |
TC1400000609.mm.1 |
MTA TC |