Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnlz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139504.1 | NP_001132976.1 | 2 - 3 | 487 | 114 | |
NM_026828.3 | NP_081104.1 | 2 - 3 | 483 | 114 | ||
GenBank mRNA | AK004089.1 | - | 2 - 3 | 473 | 114 | |
AK142070.1 | - | 2 - 3 | 484 | 114 | ||
BC139333.1 | - | 2 - 3 | 447 | 8862355 | 114 | |
BC139334.1 | - | 2 - 3 | 446 | 8862356 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |