Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172455.3 | NP_766043.3 | 8 - 9 | 1054 | 94 | |
GenBank mRNA | AK045663.1 | - | 6 - 7 | 804 | 94 | |
DQ061860.1 | - | 8 - 9 | 1055 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |