Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_001160239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_001160239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_001190436.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_001190436.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_007990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
NM_007990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001117.mm.1 |
TC1900000096.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001114.mm.1 |
NM_001160239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001114.mm.1 |
NM_001160239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001114.mm.1 |
NM_007990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001114.mm.1 |
NM_007990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001114.mm.1 |
TC1900000096.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_001160239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_001160239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_001190436.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_001190436.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_007990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
NM_007990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001120.mm.1 |
TC1900000096.mm.1 |
MTA TC |