Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008630.2 | NP_032656.1 | 2 - 3 | 324 | 91 | |
GenBank mRNA | AK002567.1 | - | 2 - 3 | 159 | 91 | |
BC031758.1 | - | 2 - 3 | 141 | 2651471 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |