Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011118.2 | NP_035248.2 | 3 - 4 | 375 | 97 | |
GenBank mRNA | AK145995.1 | - | 3 - 4 | 377 | 97 | |
AK164068.1 | - | 3 - 4 | 376 | 97 | ||
BC132078.1 | - | 3 - 4 | 357 | 40130355 | 97 | |
BC132080.1 | - | 3 - 4 | 357 | 40130357 | 97 |
Gene Symbol: Prl2c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011954.2 | NP_036084.2 | 365 | 97 | ||
GenBank mRNA | BC058816.1 | - | 365 | 30053850 | 97 | |
BC064772.1 | - | 356 | 30434146 | 97 | ||
BC100299.1 | - | 347 | 30713114 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |