Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025407.2 | NP_079683.2 | 10 - 11 | 1286 | 84 | |
GenBank mRNA | AK002292.1 | - | 10 - 11 | 1223 | 84 | |
AK004031.1 | - | 10 - 11 | 1232 | 84 | ||
AK010553.1 | - | 10 - 11 | 1226 | 84 | ||
AK013128.1 | - | 10 - 11 | 1226 | 84 | ||
AK151764.1 | - | 10 - 11 | 1228 | 84 | ||
AK160337.1 | - | 10 - 11 | 1233 | 84 | ||
AK167911.1 | - | 10 - 11 | 1226 | 84 | ||
AK168218.1 | - | 10 - 11 | 1228 | 84 | ||
AK178594.1 | - | 2 - 3 | 236 | 84 | ||
AK183917.1 | - | 2 - 3 | 237 | 84 | ||
AK214273.1 | - | 2 - 3 | 236 | 84 | ||
BC018405.1 | - | 3 - 4 | 339 | 84 | ||
BC022926.1 | - | 3 - 4 | 1400 | 84 | ||
BC040094.1 | - | 3 - 4 | 1400 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |