Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028607.1 | NP_082883.1 | 2 - 3 | 210 | 109 | |
GenBank mRNA | AK010745.1 | - | 2 - 3 | 207 | 109 | |
AK145313.1 | - | 2 - 3 | 211 | 109 | ||
BC065806.1 | - | 2 - 3 | 203 | 1547643 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |