Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029044.2 | NP_083320.1 | 10 - 11 | 1327 | 110 | |
GenBank mRNA | AK015430.1 | - | 10 - 11 | 1329 | 110 | |
BC018412.1 | - | 10 - 11 | 1340 | 3673669 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |